LY 303511 ≥95% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX270410M005EA 581 EUR
ALEXALX270410M005 ALEXALX270410M001
LY 303511 ≥95% (by HPLC)
LY 303511

Useful negative control for the PI(3)K (phosphoinositide 3-kinase) inhibitor LY-294,002. Does not inhibit PI(3)K even at concentrations.


Yellow solid.


Soluble in DMSO.

Formula: C₁₉H₁₈N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD03453556
CAS Number: 154447-38-8
https://export.vwr.com/stibo/web/std.lang.all/75/53/45347553.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR