Rockout ≥98% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX270429M005EA 56 EUR
ALEXALX270429M005
Rockout ≥98% (by HPLC)
Rockout

A cell permeable, selective, and ATP-competitive inhibitor of Rho kinase (ROCK) activity (IC50=25µM). Does not inhibit the activation of Rho nor does it affect the in vitro activities of MLCK, PKCα, and SAPK2a/p38α. Shown to be less potent than Y-27632.


  • ROCK inhibitor


Off-white to beige solid.


Soluble in DMSO or dimethyl formamide.

Formula: C₁₃H₁₀N₂
MW: 194,2
CAS Number: 7272-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR