SANT-2 ≥95%

Supplier: Enzo Life Sciences

ALEXALX270436M001EA 335 EUR
ALEXALX270436M001
SANT-2 ≥95%
SANT-2

Smoothened inhibitor


Cell permeable antagonist of the Smoothened (Smo) protein function. Inhibitor of SAG (Prod. No. ALX-270-426) activity. Chemical tool to study the Hedgehog (Hh) signalling pathway and to explore therapeutic uses for inhibitors of Smo.

Formula: C₂₆H₂₆ClN₃O₄
MW: 480 g/mol
CAS Number: 329196-48-7
https://export.vwr.com/stibo/web/std.lang.all/75/68/45347568.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR