AGK2 ≥95%

Supplier: Enzo Life Sciences

ALEXALX270484M005EA 252 EUR
ALEXALX270484M005 ALEXALX270484M001
AGK2 ≥95%
AGK2

SIRT2 inhibitor


Potent, selective and cell permeable inhibitor of sirtuin 2 (SIRT2) (IC50=3.5μM). Rescues α-synuclein-mediated toxicity. Modifies inclusion morphology in a cellular model of Parkinson’s disease. Protects against dopaminergic cell death. Leads to an increase in acetylated tubulin.

Formula: C₂₃H₁₃Cl₂N₃O₂
MW: 434,28 g/mol
MDL Number: MFCD01909444
CAS Number: 304896-28-4
https://export.vwr.com/stibo/web/std.lang.all/75/99/45347599.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR