TCS JNK 5a ≥95% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX270490M005EA 581 EUR
ALEXALX270490M005
TCS JNK 5a ≥95% (by HPLC)
TCS JNK 5a

Thienylnaphthamide compound. Potent inhibitor of JNK2 (pIC50=6.5) and JNK3 (pIC50=6.7) targeting the ATP-binding site. Has no activity against JNK1.


  • Jnk inhibitor


Tan solid.


Soluble in DMSO (10mg/ml).

Formula: C₂₀H₁₆N₂OS
MW: 332,42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01005305
CAS Number: 312917-14-9
https://export.vwr.com/stibo/web/std.lang.all/76/00/45347600.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR