Cylindrospermopsin ≥95% (by HPLC)
Supplier: Enzo Life Sciences
ALEXALX350149C100EA 569 EUR
ALEXALX350149C100 ALEXALX350149C025 ALEXALX350149M001
Cylindrospermopsin ≥95% (by HPLC)
Cylindrospermopsin
Protein synthesis inhibitor
Tricyclic alkaloid hepatotoxin. Exhibits a completely different mechanism of toxicity than microcystins. Protein synthesis inhibitor. Might be carcinogenic. Inhibits pyrimidine nucleotide synthesis.
Isolated from Cylindrospermopsis raciborskii.
CAS Number: 143545-90-8
https://export.vwr.com/stibo/web/std.lang.all/76/89/45347689.jpg