Jasplakinolide ≥97% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX350275C050EA 352 EUR
ALEXALX350275C050 ALEXALX350275C100
Jasplakinolide ≥97% (by HPLC)
Jasplakinolide

F-actin probe


Cell permeable F-actin probe. Induces actin polymerization. Completely inhibits the binding of phalloidin to F-actin. Antifungal and anti-proliferative agent.


Isolated from Jaspis splendens.

Formula: C₃₆H₄₅BrN₄O₆
CAS Number: 102396-24-7
https://export.vwr.com/stibo/web/std.lang.all/77/29/45347729.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR