Mitoxantrone dihydrochloride (synthetic) ≥97%

Supplier: Enzo Life Sciences

Synonyms: Novantrone1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione dihydrochloride

ALEXALX400050M010EA 64.4 EUR
ALEXALX400050M010 ALEXALX400050M050
Mitoxantrone dihydrochloride (synthetic) ≥97%
Mitoxantrone dihydrochloride

Antiviral, antibacterial, antiprotozoal, immunomodulating, and antineoplastic cytostatic anthraquinone derivative. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Induces interstrand DNA cross-links and DNA-protein cross-links in cellular systems. Has been shown to be an inhibitor of DNA methylation.


  • Blue to black crystalline solid
  • Soluble in water; slightly soluble in 100% ethanol or methanol; insoluble in acetonitrile or acetone

Formula: C₂₂H₃₀Cl₂N₄O₆
MW: 517,404 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00242943
CAS Number: 70476-82-3
https://export.vwr.com/stibo/web/std.lang.all/06/08/45390608.jpg

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR