S-Nitrosoglutathione (GSNO) ≥98% (by TLC)

Supplier: Enzo Life Sciences

ALEXALX420002M100EA 450 EUR
ALEXALX420002M100 ALEXALX420002M025
S-Nitrosoglutathione (GSNO) ≥98% (by TLC)
S-Nitrosoglutathione (GSNO)

Nitric oxide donor


NO donor which spontaneously releases NO under physiological conditions. Half-life of 159 hours under physiological conditions. Selectively inhibits platelet activation and displays cardioprotective effects. Inhibits NF-kappaB activation. Inhibits endothelial cell proliferation and induces apoptosis in T cells.

Formula: C₁₀H₁₆N₄O₇S
MW: 336,319 g/mol
CAS Number: 57564-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR