Compound 48/80 trihydrochloride ≥98%
Supplier: Enzo Life Sciences
ALEXALX420008M100EA 118 EUR
ALEXALX420008M100
Compound 48/80 trihydrochloride ≥98%
Compound 48/80 trihydrochloride
G protein activator
Compound 48/80 is an oligomeric mixture of condensation products of N-methyl-p-methoxyphenethylamine and formaldehyde. It activates G proteins by a mechanism analogous to that of mastoparan. It inhibits calmodulin and human platelet PLC and exhibits a concentration dependent biphasic modulation of human platelet PLA2.
CAS Number: 94724-12-6