4EGI-1 ≥95%

Supplier: Enzo Life Sciences

ALEXALX430155M002EA 373 EUR
ALEXALX430155M002
4EGI-1 ≥95%
4EGI-1

Selective and cell permeable inhibitor of the interaction between the human eukaryotic translation initiation factors 4E (eIF4E) and 4G (eIF4G).


  • eIF inhibitor


Brown solid.


Soluble in DMSO.

Formula: C₁₈H₁₂Cl₂N₄O₄S
MW: 451,3
Storage Temperature: Freezer
CAS Number: 315706-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR