Galantamine hydrobromide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX550336M050EA 234 EUR
ALEXALX550336M050
Galantamine hydrobromide ≥98% (by HPLC)
Galantamine hydrobromide

Specific, competitive and reversible acetylcholinesterase inhibitor. Alzheimer's disease therapeutic.


  • Acetylcholine esterase inhibitor


White to off-white crystalline solid.


Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml); sparingly soluble in water.

Formula: C₁₇H₂₁NO₃·HBr
MW: 368,27 g/mol
MDL Number: MFCD00067672
CAS Number: 1953-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR