Dibutyryl-cGMP ≥96%

Supplier: Enzo Life Sciences

ALEXBML-CN215-0050EA 559 EUR
ALEXBML-CN215-0050 ALEXBML-CN215-0010
Dibutyryl-cGMP ≥96%
Dibutyryl-cGMP

A cell-permeable cGMP analog which preferentially activates cGMP-dependent protein kinase. Inhibits thrombin stimulated arachidonic acid release in human platelets. Cell permeable.


  • PKG activator


White solid.


Soluble in water (100mg/ml). Also soluble in 100% ethanol, DMSO, dimethyl formamide or methanol.

Formula: C₁₈H₂₃N₅O₉P·Na
MW: 484,4 , 23.0
Storage Temperature: Freezer
CAS Number: 51116-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR