Capsaicin ≥99% (by TLC)
Supplier: Enzo Life Sciences
ALEXBML-EI125-0200EA 171 EUR
ALEXBML-EI125-0200 ALEXBML-EI125-1000
Capsaicin ≥99% (by TLC)
Capsaicin
TRPV1 agonist
Capsaicin is an agonist at the TRPV1 receptor, a ligand-gated non-selective cation channel. Exhibits neuroprotective effects in a model of transient global cerebral ischemia. Induces release of substance P from afferent nociceptive specific neurons. Constituent of cayenne pepper. Powerful excitant of peropheral sensory nerve endings, specifically unmyelinated afferent neurons (C-fibers). Reversibly inhibits aggregation of platelets.
Isolated from Capsicum  fruit.
https://export.vwr.com/stibo/web/std.lang.all/36/28/45403628.jpg