4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione ≥95% (by TLC)

Supplier: Enzo Life Sciences

ALEXBML-EI399-0050EA 521 EUR
ALEXBML-EI399-0050 ALEXBML-EI399-0010
4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione ≥95% (by TLC)
4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione

Novel inhibitor of ubiquitin C-terminal hydrolase L3 (IC50 = 0.6µM). Displays 125-fold greater selectivity over UCH-L1 (IC50 = 75µM).


  • UCH-L3 inhibitor


Orange solid.


Soluble in DMSO (25mg/ml).

Formula: C₉H₂Cl₄O₂
MW: 283,93 g/mol
MDL Number: MFCD00239209
CAS Number: 30675-13-9
https://export.vwr.com/stibo/web/std.lang.all/38/24/45403824.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR