Methanandamide ≥98% (by TLC)
Supplier: Enzo Life Sciences
ALEXBML-FA021-0025EA
283
EUR
ALEXBML-FA021-0025
ALEXBML-FA021-0005
Methanandamide ≥98% (by TLC)
Methanandamide
Cannabinoid receptor ligand
Amidase resistant cannabinoid receptor (CB) agonist (CB1: Ki=20nM; CB2: Ki=815nM). The most potent of the series of methyl-anandamides. About 4-fold higher binding affinity for cannabinoid receptor CB1 than anandamide (Prod. No. BML-FA017) in the presence of PMSF. Does also bind to TRPV1 (Ki=4.67µM). Inhibits motor behavior resembling the effectts of THC rather than anadamide.
Formula:
C₂₃H₃₉NO₂ MW: 361,57 g/mol |
MDL Number:
MFCD00467914 CAS Number: 157182-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...