B581 ≥96%

Supplier: Enzo Life Sciences

ALEXBML-G235-0005EA 934 EUR
ALEXBML-G235-0005 ALEXBML-G235-0001
B581 ≥96%
B581

Farnesyltransferase inhibitor


A potent and selective, cell-permeable farnesyl transferase inhibitor IC50=21 nM, 30-fold selective over geranylgeranyl transferase.

Formula: C₂₂H₃₈N₄O₃S₂
MW: 470,691 g/mol
MDL Number: MFCD18910841
CAS Number: 149759-96-6
https://export.vwr.com/stibo/web/std.lang.all/38/84/45403884.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR