Z-Pro-Leu-Gly-NHOH, Enzo Life Sciences
Supplier: Enzo Life Sciences
ALEXBML-PI139-0100EA 85.4 EUR
ALEXBML-PI139-0100
Z-Pro-Leu-Gly-NHOH, Enzo Life Sciences
Proteins and Peptides
MMP inhibitor
A short peptide hydroxamate that inhibits MMPs, astacin, meprin a, and likely other metzincin family members. Can also be used for purification of collagenase.