SC-51322

Supplier: Enzo Life Sciences

ALEXBML-RA112-0005EA 124 EUR
ALEXBML-RA112-0005 ALEXBML-RA112-0025
SC-51322
SC-51322

Prostanoid receptor inhibitor


A potent and selective prostaglandin E2 antagonist (pA2=8.1). Displays in vivo analgesic activity in the mouse (ED50=0.9 mg/kg).

Formula: C₂₂H₂₀ClN₃O₄S
MDL Number: MFCD00916246
CAS Number: 146032-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR