N-(2-Hydroxyethyl)oleamide ≥98% (by TLC)
Supplier: Enzo Life Sciences
ALEXBML-SL231-0050EA
322
EUR
ALEXBML-SL231-0050
ALEXBML-SL231-0010
N-(2-Hydroxyethyl)oleamide ≥98% (by TLC)
N-(2-Hydroxyethyl)oleamide
Activates TRPV1. Endogenous anandamide-like lipid which regulates appetite but does not bind to cannabinoid receptors. In rats, food intake is inhibited by oral oleylethanolamide. Induces satiety by activation of PPARα. Inhibits ceramidase.
- TRPV1 activator
White solid.
Soluble in DMSO (>25mg/ml), methanol or 100% ethanol (>25mg/ml).
Formula:
C₂₀H₃₉NO₂ MW: 325,54 g/mol |
MDL Number:
MFCD00045972 CAS Number: 111-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...