Ansamitocin P-3 ≥93% (by HPLC)

Supplier: Enzo Life Sciences

ALEXBML-T121-0001EA 577 EUR
ALEXBML-T121-0001
Ansamitocin P-3 ≥93% (by HPLC)
Ansamitocin P-3

Ansamitocin P-3 is a maytansine analog which displays potent cytotoxicity against the human solid tumor cell lines A-549 and HT-29. It shares the same binding site as rhizoxin and phomopsin A on porcine brain tubulin. Maytansine and its analogs cause extensive disassembly of the microtubule and totally prevent tubulin spiralization.


  • Microtubule inhibitor


White to off-white crystalline solid.


Soluble in DMSO.

Formula: C₃₂H₄₃ClN₂O₉
MW: 635,1 g/mol
Storage Temperature: Freezer
CAS Number: 66547-09-9
https://export.vwr.com/stibo/web/std.lang.all/15/57/45401557.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR