PNU-74654 ≥98% (by HPLC)
Supplier: Enzo Life Sciences
ALEXBML-WN101-0025EA 483 EUR
ALEXBML-WN101-0025 ALEXBML-WN101-0005
PNU-74654 ≥98% (by HPLC)
PNU-74654
Catenin blocker
Anti-tumor agent that disrupts the canonical Wnt/β-catenin pathway via blockage of the protein-protein interaction between β-catenin and TCF.
CAS Number: 113906-27-7