Dorsmorphin dihydrochloride ≥99% (by HPLC)

Supplier: Enzo Life Sciences

ALEXENZCHM1410010EA 275 EUR
ALEXENZCHM1410010 ALEXENZCHM1410005
Dorsmorphin dihydrochloride ≥99% (by HPLC)
Dorsmorphin dihydrochloride

Potent and selective AMPK inhibitor.


Potent and selective AMPK inhibitor. Dorsomorphin also inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK-2), BMPR-IA (ALK-3), and BMPR-IB (ALK-6).

MW: 472,4 g/mol
MDL Number: MFCD11112197
CAS Number: 1219168-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR