WCM (wash medium)
Supplier: Enzo Life Sciences
ALEXENZ-70002-0200EA 81.2 EUR
ALEXENZ-70002-0200
WCM (wash medium)
Cell Culture Media
For washing of spleen cells and myeloma fusion partner to remove all serum prior to the fusion process and to wash cells during fluorescent labeling.
Iscove's Modified Dulbecco's Medium based medium containing antibiotics and antimycotics