Indium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL326623-1.8GEA
188
EUR
SIAL326623-1.8G
Indium, Sigma-Aldrich®
Indium
Formula:
In MW: 114,82 g/mol Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 156 °C Density: 7,3 g/cm³ (20 °C) |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...