Indium, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL340863-13GEA 371 EUR
SIAL340863-13G
Indium, Sigma-Aldrich®
Indium
Formula: In
MW: 114,82 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7,3 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR