Indium 99.999% (trace metals basis), powder

Supplier: Thermo Scientific

ACRO193180250EA 635 EUR
ACRO193180250 ACRO193181000 ACRO193180050
Indium 99.999% (trace metals basis), powder
Indium
Formula: In
MW: 114,82 g/mol
Boiling Pt: 2000 °C (760 mmHg)
Melting Pt: 156 °C
Density: 7,3 g/cm³
MDL Number: MFCD00134048
CAS Number: 7440-74-6
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤0.001 %
Anticaking agent 2 % (MgO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR