Graphite ≥99.9%, tape, square weave, Width: 100 mm (3.94 in), Thickness: 0.56 mm (0.022 in)

10136.BYEA 329 EUR
10136.BY
Graphite ≥99.9%, tape, square weave, Width: 100 mm (3.94 in), Thickness: 0.56 mm (0.022 in)
Graphite
Formula: C
MW: 12,01 g/mol
Melting Pt: 3827 °C
Density: 2,09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
Merck Index: 12,04560

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR