Gold, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL326542-500MGEA
157
EUR
SIAL326542-500MG
SIAL326542-5G
Gold, Sigma-Aldrich®
Gold
Formula:
Au MW: 196,97 g/mol Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19,3 g/cm³ (20 °C) |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...