Gold, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL373168-6.4GEA 1580 EUR
SIAL373168-6.4G
Gold, Sigma-Aldrich®
Gold
Formula: Au
MW: 196,97 g/mol
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19,3 g/cm³ (20 °C)
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR