Copper, Sigma-Aldrich®

Supplier: Merck

SIAL365327-21.5GEA 215 EUR
SIAL365327-21.5G
Copper, Sigma-Aldrich®
Copper
Formula: Cu
MW: 63,55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,94 g/cm³ (20 °C)
MDL Number: MFCD00010965
CAS Number: 7440-50-8
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR