Tungsten ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)

Supplier: Thermo Scientific

13980.FYEA 577 EUR
13980.FY 13980.FG
Tungsten ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Tungsten
Formula: W
MW: 183,84 g/mol
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR