Iron ≥99.5% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)

44687.GTEA 329 EUR
44687.GT
Iron ≥99.5% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)
Iron
Formula: Fe
MW: 55,85 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7,86 g/cm³ (20 °C)
MDL Number: MFCD00010999
CAS Number: 7439-89-6
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR