Palladium ≥99.9% (trace metals basis), foil, Thickness: 0.05 mm, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL411450-1.5GEA
831
EUR
SIAL411450-1.5G
SIAL411450-375MG
Palladium ≥99.9% (trace metals basis), foil, Thickness: 0.05 mm, Sigma-Aldrich®
Palladium
Formula:
Pd MW: 106,420 g/mol Boiling Pt: 3167 °C (1013 hPa) Melting Pt: 1555 °C Density: 12,02 g/cm³ (20 °C) |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 Merck Index: 12,07121 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...