Platinum, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL267155-1GEA 392 EUR
SIAL267155-1G SIAL267155-5G
Platinum, Sigma-Aldrich®
Platinum
Formula: Pt
MW: 195,08 g/mol
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21,447 g/cm³ (20 °C)
MDL Number: MFCD00011179
CAS Number: 7440-06-4
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR