Petroleum spirit, 35…60 °C, PHOTREX® for spectrophotometry, J.T.Baker®

9270-03EA 0 EUR
9270-03
Petroleum spirit, 35…60 °C, PHOTREX® for spectrophotometry, J.T.Baker®
Petroleum spirit
Boiling Pt: 35…60 °C (1013 hPa)
MDL Number: MFCD00081849
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Acidity Passes Test
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤1.0
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 270 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm ≤0.010
Boiling Range(Lower) 35 - 60 °C
Boiling Range(Upper) 35 - 60 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR