Agarose, Sigma-Aldrich®

Supplier: ROCHE

ROCH11816594001EA 2800 EUR
ROCH11816594001 ROCH1816594
Agarose, Sigma-Aldrich®
Agarose
Melting Pt: 60…90 °C
MDL Number: MFCD00081294
CAS Number: 9012-36-6
https://export.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR