Duo-iODN (inhibitory ODN) (synthetic) endotoxin-free (Bulk)
Supplier: Enzo Life Sciences
ALEXALX746355M001EA 609 EUR
ALEXALX746355M001
Duo-iODN (inhibitory ODN) (synthetic) endotoxin-free (Bulk)
Duo-iODN (inhibitory ODN)
Inhibitor of TLR7 and TLR9
Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Sequence of an inhibitory ODN for in vivo use in rodents (50-150µg per injection): prototype class I that potently inhibits TLR9 and TLR7 signaling.
Endotoxin content: <0,0002 EU/µg (LAL test; BioWhittaker)