Chain tube racks, VWR®
Supplier: VWR, part of Avantor
VWR®
479-2129EA 38.8 EUR
479-2129
Chain tube racks, VWR®
Racks Tube Racks
Plastic, coloured.
  • Chain tube rack can be modified to be used in multiple applications including in water or ice baths, in phlebotomy trays, in a Duraporter® specimen transport case, or anywhere a tube rack is needed
  • Maximise valuable bench space with the flexibility to hold multiple size tubes (5/15 ml, 15 to 17 mm tubes) in one configurable rack
  • Open design allows exposure to water and ice baths helping to reduce identification errors
  • Create different shapes and rack sizes with up to 16 wells, depending on the need: Straight line, circle, 2×8, 4×4, etc.
  • Link together multiple chain rack packs to accommodate larger tube rack configuration requirements
  • Assorted colours for easy identification of tubes or testing (blue, green, yellow and natural)
  • Not autoclavable
The chain tube rack has a unique linkable design that enables multiple configurations to form different shaped tube racks. Each well has the flexibility to hold 5 ml tubes on one side, or rotate it 180° to hold 15 ml or 15 to 17 mm tubes (round and conical bottom). Mix and match the well sizes to create one rack that holds multiple sized tubes in a variety of functional configurations.