Hydrochloric acid 0.1 mol/l (0.1 N), Supelco®


M-Clarity™ quality level = MQ100

SUPL2104-50MLEA 111 EUR
SUPL2104-50ML
Hydrochloric acid 0.1 mol/l (0.1 N), Supelco®
Hydrochloric acid
Formula: HCl
MW: 36,46 g/mol
Density: 1,004 g/cm³ (25 °C)
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR