Sodium hydroxide 25% (w/v) in aqueous solution

43771.K2EA 32.5 EUR
43771.K2
Sodium hydroxide 25% (w/v) in aqueous solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR