25-Hydroxyvitamin D₂ ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

ALEXBML-DM101-0001EA 282 EUR
ALEXBML-DM101-0001
25-Hydroxyvitamin D₂ ≥98% (by HPLC)
25-Hydroxyvitamin D₂

High purity vitamin D₂ intermediate.


25-Hydroxyvitamin D₂ is an intermediate produced from vitamin D₂ by cytochrome P450 isoforms in serum. This primary metabolite of vitamin D₂ can be used to indirectly measure the concencentration of vitamin D via HPLC and UV.


Soluble in DMSO, ethanol or chloroform.

Formula: C₂₈H₄₄O₂
MW: 412,65 g/mol
MDL Number: MFCD02683431
CAS Number: 21343-40-8
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR