Lysates, A431 (human)
Supplier: Enzo Life Sciences
ALEXADILYCA4102FEA 132 EUR
ALEXADILYCA4102F ALEXADILYCA4100F
Lysates, A431 (human)
Cells
The A431 cell line is derived from a human epithelial carcinoma and is commonly used as a positive control for the study of growth factor action and binding due to its high expression of EGFR.
Dilution range: 20 µg/lane (Western blotting)
Liquid lysates in SDS-PAGE sample buffer, pH 6,8, heated for 10 minutes at 70 °C and containing Tris-HCl (40 mM), SDS (1%), DTT (50 mM), glycerol (7,5%), and bromophenol blue (0,003%).