DNA (Deoxyribonucleic acid) (from calf thymus), purified

Supplier: Enzo Life Sciences

ALEXALX840040C010EA 136 EUR
ALEXALX840040C010
DNA (Deoxyribonucleic acid) (from calf thymus), purified
DNA (Deoxyribonucleic acid)

Purified from calf thymus and activated by digestion with DNAse I.


For use with PARP-1 or PARP-2 in vitro activity tests


Concentration: 0,2 mg/ml

Storage Temperature: Refrigerator
MDL Number: MFCD00130922
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR