Procaspase-3-activator (PAC-1) ≥98% (by HPLC), white to off-white solid

Supplier: Enzo Life Sciences
Warning

ALEXALX430146M050EA 348 EUR
ALEXALX430146M050 ALEXALX430146M010
Procaspase-3-activator (PAC-1) ≥98% (by HPLC), white to off-white solid
Procaspase-3-activator (PAC1, (4-benzylpiperazino)acetic acid-(3-allyl-2-hydroxybenzylidene)hydrazide)

Caspase-3 activator


First molecule to directly activate procaspase-3 to caspase-3 in vitro. Has been shown to induce apoptosis in cancer cells.


Soluble in acetonitrile, DMSO, dioxane or methanol.

Formula: C₂₃H₂₈N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD03302441
CAS Number: 315183-21-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Purity (HPLC (UV)) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR