Magnesium sulphate heptahydrate, Electran for molecular biology

437044KEA 0 EUR
437044K
Magnesium sulphate heptahydrate, Electran for molecular biology
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246,48 g/mol
Melting Pt: 1124 °C
Density: 2,66 g/cm³ (20 °C)
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR