DIPPMPO ≥98% (by TLC, 1H-NMR, 13C-NMR)
Supplier: Enzo Life Sciences
ALEXALX430119M050EA
364
EUR
ALEXALX430119M050
DIPPMPO ≥98% (by TLC, 1H-NMR, 13C-NMR)
DIPPMPO
Most efficient spin trap for the in vitro and in vivo detection of O-, N-, S-, and C-centered free radicals.
DIPPMPO has a longer life-time than DMPO. More lipophilic (Kp=2.1) than DEPMPO.
Very hygroscopic and soluble in water, 100% ethanol or methylene chloride.
Formula:
C₁₁H₂₂NO₄P MW: 263,27 g/mol Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
MDL Number:
MFCD08702711 CAS Number: 527704-58-1 |
Specification Test Results
Purity (TLC, ¹H- and ¹³C-NMR) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...