NBQX ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Warning

ALEXALX550044M005EA 157 EUR
ALEXALX550044M005 ALEXALX550044M025
NBQX ≥99% (by HPLC)
NBQX

Most potent and selective AMPA/kainate receptor antagonist. More soluble form is NBQX disodium salt.


Soluble in DMSO. A 1 µM aqueous solution may be prepared by taking up in >2.5 equivalents of NaOH and backtitrating to pH 8.0 to 8.5 with concentrated HCl.

Formula: C₁₂H₈N₄O₆S
MW: 336,3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00270054
CAS Number: 118876-58-7
https://export.vwr.com/stibo/web/std.lang.all/07/96/45390796.jpg

Order Now

Specification Test Results

Purity (HPLC) ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR