24(S),25-Epoxycholesterol ≥98% (by TLC)

Supplier: Enzo Life Sciences


Packaged under inert gas.

ALEXBML-GR231-0005EA 1290 EUR
ALEXBML-GR231-0005 ALEXBML-GR231-0001
24(S),25-Epoxycholesterol ≥98% (by TLC)
24(S),25-Epoxycholesterol

LXR ligand.


Endogenous agonist for the nuclear receptor LXR.


Soluble in ethanol (10 mg/ml) or DMSO (10 mg/ml).


Caution: Should be used immediately following dissolution. Store with desiccant.

Formula: C₂₇H₄₄O₂
Melting Pt: 156…159 °C
CAS Number: 77058-74-3

Order Now

Specification Test Results

Purity ≥98% (TLC)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR