Prima-1 ≥98% (by TLC), white to off-white solid

Supplier: Enzo Life Sciences
Warning

ALEXBML-GR333-0005EA 93.8 EUR
ALEXBML-GR333-0005
Prima-1 ≥98% (by TLC), white to off-white solid
Prima-1

p53 activator.


Novel cell-permeable p53 reactivating agent. Restores sequence-specific DNA binding and the active conformation to mutant p53 (in 13 of 14 p53 mutants tested) in vitro and in living cells. Induces apoptosis in human tumor cell lines and displays antitumor activity in mice.


Soluble at 25 mg/ml in ethanol, 100% DMSO and DMF. Soluble at 10 mg/ml in water.


Caution: Product is not sterile.

Formula: C₉H₁₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD04974196
CAS Number: 5608-24-2

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR